This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05000270_m1
See other ALG10B GE Assays ›
Gene Symbol:
ALG10B
Gene Name:
ALG10B, alpha-1,2-glucosyltransferase
Gene Aliases:
ALG10, KCR1
Chromosome Location:
Chr.12: 38316679 - 38329726 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05000270_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALG10B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308340.1 NP_001295269.1 2 - 3 507 137
GenBank mRNA AL712776.1 - 2 - 3 500 137

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
dolichol-linked oligosaccharide biosynthetic process
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800007323.hg.1 ENST00000476186.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007323.hg.1 ENST00000476186.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000548240.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000548240.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000548240.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000548240.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000548240.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000548240.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000551464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000551464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000551464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000551464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000551464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000551464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Clariom D Assay TC0800007323.hg.1 ENST00000476186.2 Ensembl
Affymetrix Human Clariom D Assay TC0800007323.hg.1 ENST00000476186.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000308742.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000548240.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000548240.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000548240.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000548240.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000548240.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000548240.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000551464.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000551464.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000551464.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000551464.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000551464.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000551464.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 ENST00000553138.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001013620.3 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001308340.1 Refseq
Affymetrix Human Clariom D Assay TC1200007326.hg.1 NM_001308340.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0