This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05000133_m1
See other DAO GE Assays ›
Gene Symbol:
DAO
Gene Name:
D-amino acid oxidase
Gene Aliases:
DAAO, DAMOX, OXDA
Chromosome Location:
Chr.12: 108880073 - 108901043 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05000133_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DAO
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AX776480.1 - 1 - 2 105 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AX776480.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

oxidase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000546552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000546552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000546552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000546552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000546552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000228476.7 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000546552.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000546552.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000546552.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000546552.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000546552.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547122.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547166.1 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000547768.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000548052.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000549215.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 ENST00000551281.5 Ensembl
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq
Affymetrix Human Clariom D Assay TC1200008789.hg.1 NM_001917.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0