This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04998698_m1
See other GJB1 GE Assays ›
Gene Symbol:
GJB1
Gene Name:
gap junction protein beta 1
Gene Aliases:
CMTX, CMTX1, CX32
Chromosome Location:
Chr.X: 71215212 - 71225215 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04998698_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011530907.2 XP_011529209.1 1 - 2 182 109

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011530907.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

transport
cell-cell signaling
nervous system development
purine ribonucleotide transport
gap junction assembly
protein oligomerization
transmembrane transport
gap junction channel activity
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000361726.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374022.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000374029.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 ENST00000447581.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_000166.5 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_001097642.2 Refseq
Affymetrix Human Clariom D Assay TC0X00007559.hg.1 NM_001097642.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0