This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04998236_s1
See other SHB GE Assays ›
Gene Symbol:
SHB
Gene Name:
SH2 domain containing adaptor protein B
Gene Aliases:
bA3J10.2
Chromosome Location:
Chr.9: 37915898 - 38069213 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04998236_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SHB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003028.2 NP_003019.2 1 773 82
GenBank mRNA BC029953.1 - 1 278 82
BC136581.1 - 1 646 9020571 82
BC136582.1 - 1 646 9020572 82
X75342.1 - 1 776 82

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SHB

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
apoptotic process
signal transduction
positive regulation of signal transduction
cell differentiation
vascular endothelial growth factor receptor signaling pathway
SH3/SH2 adaptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Clariom D Assay TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Clariom D Assay TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Clariom D Assay TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Clariom D Assay TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Clariom D Assay TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Clariom D Assay TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Clariom D Assay TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Clariom D Assay TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Clariom D Assay TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Clariom D Assay TC0900012231.hg.1 ENST00000377707.3 Ensembl
Affymetrix Human Clariom D Assay TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Clariom D Assay TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Clariom D Assay TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Clariom D Assay TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Clariom D Assay TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Clariom D Assay TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Clariom D Assay TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Clariom D Assay TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Clariom D Assay TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Clariom D Assay TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Clariom D Assay TC0900012231.hg.1 NM_003028.2 Refseq
Affymetrix Human Clariom D Assay TC0900012231.hg.1 NM_003028.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0