This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04996728_m1
See other RNF20 GE Assays ›
Gene Symbol:
RNF20
Gene Name:
ring finger protein 20
Gene Aliases:
BRE1, BRE1A, hBRE1
Chromosome Location:
Chr.9: 101533849 - 101563344 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04996728_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RNF20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011518862.1 XP_011517164.1 1 - 2 345 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011518862.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
regulation of mitotic cell cycle
histone monoubiquitination
negative regulation of cell migration
positive regulation of histone methylation
histone H2B ubiquitination
positive regulation of transcription, DNA-templated
negative regulation of mRNA polyadenylation
positive regulation of histone H2B ubiquitination
p53 binding
chromatin binding
transcription coactivator activity
mRNA 3'-UTR binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
histone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000374819.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000374819.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000374819.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000374819.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000374819.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000374819.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000466817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000466817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000466817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000466817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000466817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000466817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000478072.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000478072.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000478072.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000478072.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000479306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000479306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000479306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000479306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000479306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000481046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000481046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000481046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000481046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 ENST00000481046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000374819.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000374819.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000374819.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000374819.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000374819.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000374819.6 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000389120.7 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000466817.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000466817.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000466817.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000466817.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000466817.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000466817.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000478072.2 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000478072.2 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000478072.2 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000478072.2 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000479306.5 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000479306.5 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000479306.5 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000479306.5 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000479306.5 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000481046.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000481046.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000481046.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000481046.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 ENST00000481046.1 Ensembl
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq
Affymetrix Human Clariom D Assay TC0900008257.hg.1 NM_019592.6 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0