This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04995644_s1
See other GPIHBP1 GE Assays ›
Gene Symbol:
GPIHBP1
Gene Name:
glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1
Gene Aliases:
GPI-HBP1, HYPL1D
Chromosome Location:
Chr.8: 143213193 - 143217170 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04995644_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPIHBP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178172.5 NP_835466.2 4 2075 121
GenBank mRNA AF088057.1 - 1 320 121
AY245915.1 - 4 2075 121
BC035810.1 - 4 2075 5754421 121
BC063857.1 - 4 2003 6183386 121

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPIHBP1

Gene Ontology Categories:

Function(s) Process(es)

lipid transport
intracellular protein transport
protein import
protein localization to cell surface
cholesterol homeostasis
transcytosis
protein stabilization
positive regulation of lipoprotein lipase activity
triglyceride homeostasis
response to heparin
protein transmembrane transport
positive regulation of chylomicron remnant clearance
high-density lipoprotein particle binding
lipid binding
protein transmembrane transporter activity
lipase binding
chylomicron binding
lipoprotein particle binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_001301772.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_001301772.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_001301772.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_001301772.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_001301772.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_001301772.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009186.hg.1 ENST00000622500.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_001301772.1 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_001301772.1 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_001301772.1 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_001301772.1 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_001301772.1 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_001301772.1 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_178172.5 Refseq
Affymetrix Human Clariom D Assay TC0800009186.hg.1 NM_178172.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0