This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04995279_s1
See other DOCK5 GE Assays ›
Gene Symbol:
DOCK5
Gene Name:
dedicator of cytokinesis 5
Gene Aliases:
-
Chromosome Location:
Chr.8: 25184723 - 25415716 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04995279_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DOCK5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024940.7 NP_079216.4 52 9932 142
XM_011544661.1 XP_011542963.1 52 9890 142
XM_011544662.1 XP_011542964.1 51 9786 142
XM_017013865.1 XP_016869354.1 52 9742 142
XM_017013866.1 XP_016869355.1 50 10847 142
XM_017013867.1 XP_016869356.1 45 9280 142
XM_017013868.1 XP_016869357.1 46 9203 142
GenBank mRNA AK129574.1 - 2 1541 142
BF195734.1 - 1 142
CR627414.1 - 8 5180 142

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DOCK5

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
positive regulation of epithelial cell migration
positive regulation of GTPase activity
positive regulation of substrate adhesion-dependent cell spreading
negative regulation of vascular smooth muscle contraction
positive regulation of vascular associated smooth muscle cell migration
guanyl-nucleotide exchange factor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0