This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04994299_g1
See other SMPD5 GE Assays ›
Gene Symbol:
SMPD5
Gene Name:
sphingomyelin phosphodiesterase 5
Gene Aliases:
MA-nSMase
Chromosome Location:
Chr.8: 144048677 - 144051220 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04994299_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMPD5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017014128.1 XP_016869617.1 4 - 5 1369 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000528912.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009233.hg.1 ENST00000561181.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0