This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04993313_m1
See other LOC100507156 GE Assays ›
Gene Symbol:
LOC100507156
Gene Name:
uncharacterized LOC100507156
Gene Aliases:
-
Chromosome Location:
Chr.8: 23336208 - 23366125 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04993313_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100507156
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038323.1 1 - 2 1252 103
GenBank mRNA AL833246.1 - 1 - 2 1252 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800007019.hg.1 ENST00000517420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007019.hg.1 ENST00000517420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007019.hg.1 ENST00000517420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007019.hg.1 ENST00000517420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007019.hg.1 ENST00000519692.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007019.hg.1 ENST00000519692.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007019.hg.1 ENST00000519692.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800007019.hg.1 NR_038323.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007019.hg.1 NR_038323.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800007019.hg.1 NR_038323.1 Refseq
Affymetrix Human Clariom D Assay TC0800007019.hg.1 ENST00000517420.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007019.hg.1 ENST00000517420.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007019.hg.1 ENST00000517420.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007019.hg.1 ENST00000517420.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007019.hg.1 ENST00000519692.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007019.hg.1 ENST00000519692.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007019.hg.1 ENST00000519692.1 Ensembl
Affymetrix Human Clariom D Assay TC0800007019.hg.1 NR_038323.1 Refseq
Affymetrix Human Clariom D Assay TC0800007019.hg.1 NR_038323.1 Refseq
Affymetrix Human Clariom D Assay TC0800007019.hg.1 NR_038323.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0