This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04991344_s1
See other TRAF6 GE Assays ›
Gene Symbol:
TRAF6
Gene Name:
TNF receptor associated factor 6
Gene Aliases:
MGC:3310, RNF85
Chromosome Location:
Chr.11: 36483767 - 36510313 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04991344_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAF6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004620.3 NP_004611.1 7 7768 91
NM_145803.2 NP_665802.1 8 7861 91
GenBank mRNA AK128396.1 - 1 3518 91
BQ187062.1 - 1 401 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRAF6

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
activation of MAPK activity
protein polyubiquitination
ossification
neural tube closure
stimulatory C-type lectin receptor signaling pathway
toll-like receptor signaling pathway
positive regulation of T cell cytokine production
MyD88-dependent toll-like receptor signaling pathway
MyD88-independent toll-like receptor signaling pathway
protein complex assembly
I-kappaB kinase/NF-kappaB signaling
activation of NF-kappaB-inducing kinase activity
JNK cascade
antigen processing and presentation of exogenous peptide antigen via MHC class II
osteoclast differentiation
membrane protein intracellular domain proteolysis
positive regulation of protein ubiquitination
positive regulation of lipopolysaccharide-mediated signaling pathway
activation of protein kinase activity
positive regulation of interleukin-2 production
toll-like receptor 9 signaling pathway
Fc-epsilon receptor signaling pathway
T-helper 1 type immune response
positive regulation of T cell proliferation
odontogenesis of dentin-containing tooth
myeloid dendritic cell differentiation
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JUN kinase activity
positive regulation of interleukin-12 biosynthetic process
positive regulation of interleukin-6 biosynthetic process
bone resorption
positive regulation of osteoclast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell development
positive regulation of smooth muscle cell proliferation
T cell receptor signaling pathway
positive regulation of T cell activation
regulation of immunoglobulin secretion
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
protein autoubiquitination
nucleotide-binding oligomerization domain containing signaling pathway
interleukin-1-mediated signaling pathway
protein K63-linked ubiquitination
response to interleukin-1
cellular response to lipopolysaccharide
positive regulation of transcription regulatory region DNA binding
ubiquitin-protein transferase activity
tumor necrosis factor receptor binding
protein binding
zinc ion binding
ligase activity
protein kinase binding
mitogen-activated protein kinase kinase kinase binding
ubiquitin conjugating enzyme binding
ubiquitin protein ligase binding
thioesterase binding
identical protein binding
histone deacetylase binding
protein kinase B binding
protein N-terminus binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0