This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04991304_s1
See other CSTF3-AS1 GE Assays ›
Gene Symbol:
CSTF3-AS1
Gene Name:
CSTF3 antisense RNA 1 (head to head)
Gene Aliases:
-
Chromosome Location:
Chr.11: 33161657 - 33191598 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04991304_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CSTF3-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_034027.1 4 1438 120

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200007457.hg.1 ENST00000622034.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007457.hg.1 ENST00000622034.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0