This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04990283_m1
See other WEE2-AS1 GE Assays ›
Gene Symbol:
WEE2-AS1
Gene Name:
WEE2 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 141704338 - 141738230 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04990283_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: WEE2-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_015392.1 5 - 6 647 144
GenBank mRNA AK098171.1 - 5 - 6 647 144
BC101359.2 - 5 - 6 901 144
BC131564.1 - 3 - 4 277 144

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000465110.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000465110.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000465110.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000465110.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000465110.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000465110.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000473776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000473776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000486906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000486906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000486906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000486906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000486906.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000459753.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000462383.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000465110.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000465110.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000465110.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000465110.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000465110.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000465110.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000471512.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000473776.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000473776.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000478332.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000484172.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000486906.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000486906.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000486906.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000486906.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000486906.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000488785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 ENST00000495800.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Clariom D Assay TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Clariom D Assay TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Clariom D Assay TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Clariom D Assay TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Clariom D Assay TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Clariom D Assay TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Clariom D Assay TC0700012839.hg.1 NR_015392.1 Refseq
Affymetrix Human Clariom D Assay TC0700012839.hg.1 NR_015392.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0