This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04989831_g1
See other TCAF1 GE Assays ›
Gene Symbol:
TCAF1
Gene Name:
TRPM8 channel associated factor 1
Gene Aliases:
FAM115A
Chromosome Location:
Chr.7: 143851368 - 143902228 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04989831_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TCAF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001206938.1 NP_001193867.1 9 5259 106
NM_001206941.1 NP_001193870.1 8 4604 106
NM_014719.2 NP_055534.1 9 5193 106
XM_005250074.3 XP_005250131.1 9 5233 106
XM_005250075.2 XP_005250132.1 9 5235 106
XM_005250076.3 XP_005250133.1 9 5270 106
XM_005250077.2 XP_005250134.1 10 5399 106
XM_006716191.2 XP_006716254.1 9 5199 106
XM_006716192.3 XP_006716255.1 9 5368 106
XM_017012833.1 XP_016868322.1 9 5291 106
XM_017012834.1 XP_016868323.1 9 5261 106
XM_017012835.1 XP_016868324.1 10 5425 106
XM_017012836.1 XP_016868325.1 9 5225 106
GenBank mRNA AF119896.2 - 1 106
BC039457.1 - 1 1153 106

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF119896.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TCAF1

Gene Ontology Categories:

Function(s) Process(es)

transport
negative regulation of cell migration
positive regulation of protein targeting to membrane
positive regulation of anion channel activity
protein binding
ion channel binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0