Assay Details
Gene Symbol: UBN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173569.3 | NP_775840.3 | 3 - 4 | 654 | 133 | |
XM_005250249.3 | XP_005250306.2 | 3 - 4 | 2571 | 133 | ||
XM_011516002.2 | XP_011514304.1 | 3 - 4 | 2574 | 133 | ||
XM_011516003.1 | XP_011514305.1 | 3 - 4 | 2574 | 133 | ||
GenBank mRNA | AI803206.1 | - | 3 - 4 | 292 | 133 | |
AK125956.1 | - | 3 - 4 | 434 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000609762.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000609762.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000609762.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | ENST00000609762.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000473989.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000483726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000486663.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000609762.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000609762.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000609762.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | ENST00000609762.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009333.hg.1 | NM_173569.3 | Refseq |
