Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000473300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000473300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000473300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000473300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000496754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000496754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000496754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
ENST00000496754.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000304402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000473300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000473300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000473300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000473300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000496754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000496754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000496754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
ENST00000496754.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008753.hg.1 |
NM_005295.2 |
Refseq |