This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04987123_m1
See other COX7A2 GE Assays ›
Gene Symbol:
COX7A2
Gene Name:
cytochrome c oxidase subunit 7A2
Gene Aliases:
COX7AL, COX7AL1, COXVIIAL, COXVIIa-L, VIIAL
Chromosome Location:
Chr.6: 75237675 - 75243928 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04987123_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COX7A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029466.1 2 - 3 352 120
GenBank mRNA CB139775.1 - 2 - 3 211 120

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

hydrogen ion transmembrane transport
cytochrome-c oxidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000377978.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000377978.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000377978.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000377978.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000472311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000472311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000472311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000472311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000472311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000472311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000481061.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000481061.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000481061.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000481061.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370081.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000370089.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000377978.3 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000377978.3 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000377978.3 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000377978.3 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000459637.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000460985.1 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000472311.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000472311.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000472311.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000472311.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000472311.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000472311.6 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000481061.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000481061.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000481061.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000481061.2 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 ENST00000509698.5 Ensembl
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NM_001865.3 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NR_029466.1 Refseq
Affymetrix Human Clariom D Assay TC0600012315.hg.1 NR_029466.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm