Assay Details
Gene Symbol: ORC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK302750.1 | - | 15 - 16 | 1705 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: ORC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK302750.1 | - | 15 - 16 | 1705 | 92 |
Set Membership: |
![]() ![]() ![]() |