This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04983023_g1
See other NFATC2IP GE Assays ›
Gene Symbol:
NFATC2IP
Gene Name:
nuclear factor of activated T-cells 2 interacting protein
Gene Aliases:
ESC2, NIP45, RAD60
Chromosome Location:
Chr.16: 28950988 - 28967097 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04983023_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NFATC2IP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032815.3 NP_116204.3 6 - 7 1004 61
GenBank mRNA AF458593.1 - 6 - 7 1014 61
AK027545.1 - 3 - 4 419 61
AK074761.1 - 6 - 7 994 61
AK297333.1 - 1 - 2 349 61
AK304606.1 - 1 - 2 271 61
AK316535.1 - 1 - 2 410 61
BC018311.1 - 4 - 5 508 61
BC021551.1 - 3 - 4 387 61
BC068007.1 - 3 - 4 244 61
BC080628.1 - 6 - 7 671 61
BC101741.1 - 6 - 7 1064 8069247 61
BC112182.1 - 6 - 7 1064 8327650 61

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NFATC2IP

Gene Ontology Categories:

Function(s) Process(es)

cytokine production
protein sumoylation
positive regulation of transcription from RNA polymerase II promoter
protein tag

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700010194.hg.1 ENST00000623773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010194.hg.1 ENST00000623773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000562977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000562977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000562977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000562977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000562977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000565752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000565752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000565752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000565752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000565919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000565919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000565919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000565919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000568998.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 ENST00000578855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600007374.hg.1 NR_039742.1 Refseq
Affymetrix Human Clariom D Assay TC1700010194.hg.1 ENST00000623773.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010194.hg.1 ENST00000623773.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000320805.8 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000562977.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000562977.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000562977.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000562977.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000562977.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000564978.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000565752.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000565752.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000565752.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000565752.5 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000565919.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000565919.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000565919.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000565919.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568148.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568998.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568998.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568998.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000568998.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 ENST00000578855.1 Ensembl
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NM_032815.3 Refseq
Affymetrix Human Clariom D Assay TC1600007374.hg.1 NR_039742.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0