This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04980370_m1
See other MIR100HG GE Assays ›
Gene Symbol:
MIR100HG
Gene Name:
mir-100-let-7a-2 cluster host gene
Gene Aliases:
AGD1, linc-NeD125, lncRNA-N2
Chromosome Location:
Chr.11: 122089103 - 122203062 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04980370_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR100HG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK311733.1 - 2 - 3 790 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000524376.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000524376.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000524376.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000524376.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000524376.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000524376.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000524376.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000524376.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000524376.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000524376.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000526674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000526674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000527474.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000528986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529804.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529804.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529804.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529804.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529804.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529804.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000529823.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000530072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000530072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000530072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000530072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000530072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000530072.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000530955.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000530955.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531381.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531470.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531470.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531470.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531470.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000531629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532315.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532315.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532315.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532315.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532319.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532350.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532350.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532350.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532350.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532350.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532350.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532350.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532350.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532350.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532350.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532350.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532350.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532890.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532890.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532890.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532890.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532890.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532890.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532890.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532890.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532890.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000532890.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000533109.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000533109.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000533109.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000533109.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534195.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534195.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534195.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534195.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534195.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534297.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534496.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534782.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534782.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534782.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534782.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534782.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534782.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534782.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534782.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534782.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000534782.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000560104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000560104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000560104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000560104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000560104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 ENST00000560104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NM_001001786.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NM_001001786.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NM_001001786.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NM_001001786.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NM_001001786.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NM_001001786.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012583.hg.1 NR_024430.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm