This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04979945_s1
See other F2RL1 GE Assays ›
Gene Symbol:
F2RL1
Gene Name:
F2R like trypsin receptor 1
Gene Aliases:
GPR11, PAR2
Chromosome Location:
Chr.5: 76819008 - 76835315 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04979945_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: F2RL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC018130.1 - 1 75 3895653 71
DB463838.1 - 1 127 71
U34038.1 - 1 68 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC018130.1, U34038.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein coupled receptor

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 ENST00000296677.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 ENST00000296677.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 ENST00000296677.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 ENST00000296677.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 ENST00000296677.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 ENST00000296677.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 ENST00000514165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 ENST00000514165.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 NM_005242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 NM_005242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 NM_005242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 NM_005242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 NM_005242.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007834.hg.1 NM_005242.5 Refseq
Affymetrix Human Clariom D Assay TC0500007834.hg.1 ENST00000296677.4 Ensembl
Affymetrix Human Clariom D Assay TC0500007834.hg.1 ENST00000296677.4 Ensembl
Affymetrix Human Clariom D Assay TC0500007834.hg.1 ENST00000296677.4 Ensembl
Affymetrix Human Clariom D Assay TC0500007834.hg.1 ENST00000296677.4 Ensembl
Affymetrix Human Clariom D Assay TC0500007834.hg.1 ENST00000296677.4 Ensembl
Affymetrix Human Clariom D Assay TC0500007834.hg.1 ENST00000296677.4 Ensembl
Affymetrix Human Clariom D Assay TC0500007834.hg.1 ENST00000514165.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007834.hg.1 ENST00000514165.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007834.hg.1 NM_005242.5 Refseq
Affymetrix Human Clariom D Assay TC0500007834.hg.1 NM_005242.5 Refseq
Affymetrix Human Clariom D Assay TC0500007834.hg.1 NM_005242.5 Refseq
Affymetrix Human Clariom D Assay TC0500007834.hg.1 NM_005242.5 Refseq
Affymetrix Human Clariom D Assay TC0500007834.hg.1 NM_005242.5 Refseq
Affymetrix Human Clariom D Assay TC0500007834.hg.1 NM_005242.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0