This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04977205_m1
See other ICE1 GE Assays ›
Gene Symbol:
ICE1
Gene Name:
interactor of little elongation complex ELL subunit 1
Gene Aliases:
KIAA0947
Chromosome Location:
Chr.5: 5422673 - 5490234 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04977205_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ICE1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015325.2 NP_056140.1 7 - 8 668 88
XM_011513999.2 XP_011512301.1 7 - 8 668 88
XM_017009285.1 XP_016864774.1 7 - 8 660 88
GenBank mRNA AB023164.3 - 7 - 8 647 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017009285.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Recommended Assay Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein complex assembly
snRNA transcription from RNA polymerase II promoter
snRNA transcription from RNA polymerase III promoter
positive regulation of transcription from RNA polymerase III promoter
positive regulation of intracellular protein transport
protein binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000505464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000505464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000505464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000505464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000505464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000296564.8 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000505464.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000505464.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000505464.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000505464.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000505464.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 ENST00000512608.5 Ensembl
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq
Affymetrix Human Clariom D Assay TC0500006614.hg.1 NM_015325.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0