Assay Details
Gene Symbol: ICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015325.2 | NP_056140.1 | 10 - 11 | 837 | 97 | |
XM_011513999.2 | XP_011512301.1 | 10 - 11 | 837 | 97 | ||
GenBank mRNA | AB023164.3 | - | 10 - 11 | 816 | 97 | |
CR749441.1 | - | 3 - 4 | 108 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein complex assembly
snRNA transcription from RNA polymerase II promoter
snRNA transcription from RNA polymerase III promoter
positive regulation of transcription from RNA polymerase III promoter
positive regulation of intracellular protein transport
protein binding
protein homodimerization activity
positive regulation of protein complex assembly
snRNA transcription from RNA polymerase II promoter
snRNA transcription from RNA polymerase III promoter
positive regulation of transcription from RNA polymerase III promoter
positive regulation of intracellular protein transport
protein binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000505464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000505464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000505464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000505464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000505464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000296564.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000505464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000505464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000505464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000505464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000505464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | ENST00000512608.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500006614.hg.1 | NM_015325.2 | Refseq |
