This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04975712_s1
See other LRP2BP GE Assays ›
Gene Symbol:
LRP2BP
Gene Name:
LRP2 binding protein
Gene Aliases:
-
Chromosome Location:
Chr.4: 185363878 - 185397160 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04975712_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LRP2BP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AL161975.1 - 1 1345 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000511404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000511404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000511404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000511404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 ENST00000511404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000328559.11 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000510776.5 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000511404.1 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000511404.1 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000511404.1 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000511404.1 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 ENST00000511404.1 Ensembl
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq
Affymetrix Human Clariom D Assay TC0400013001.hg.1 NM_018409.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0