This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04975093_m1
See other NPY1R GE Assays ›
Gene Symbol:
NPY1R
Gene Name:
neuropeptide Y receptor Y1
Gene Aliases:
NPY1-R, NPYR
Chromosome Location:
Chr.4: 163323962 - 163344847 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04975093_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPY1R
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005263031.3 XP_005263088.1 1 - 2 545 116

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005263031.3 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

outflow tract morphogenesis
glucose metabolic process
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
locomotory behavior
feeding behavior
blood circulation
regulation of blood pressure
sensory perception of pain
peristalsis
response to estradiol
regulation of multicellular organism growth
positive regulation of vasoconstriction
negative regulation of hormone secretion
negative regulation of cytosolic calcium ion concentration
peptide YY receptor activity
pancreatic polypeptide receptor activity
neuropeptide Y receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504790.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504790.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504790.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504790.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504790.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000504790.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000511901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000511901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000511901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000511901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000512819.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000512819.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000512819.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000512819.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000512819.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000512819.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000515701.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000515701.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000515701.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000515701.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 ENST00000515701.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000296533.2 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504391.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504790.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504790.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504790.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504790.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504790.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000504790.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000509586.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000511901.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000511901.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000511901.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000511901.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000512819.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000512819.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000512819.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000512819.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000512819.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000512819.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000515701.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000515701.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000515701.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000515701.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 ENST00000515701.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq
Affymetrix Human Clariom D Assay TC0400012305.hg.1 NM_000909.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0