This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04974975_m1
See other GRID2 GE Assays ›
Gene Symbol:
GRID2
Gene Name:
glutamate ionotropic receptor delta type subunit 2
Gene Aliases:
GluD2, SCAR18
Chromosome Location:
Chr.4: 92304249 - 93810330 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04974975_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GRID2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011531893.2 XP_011530195.2 7 - 8 7691 127
XM_011531894.2 XP_011530196.1 5 - 6 730 127
XM_011531895.2 XP_011530197.1 5 - 6 647 127
XM_017008119.1 XP_016863608.1 7 - 8 1228 127
XM_017008120.1 XP_016863609.1 7 - 8 7691 127
XM_017008121.1 XP_016863610.1 7 - 8 7691 127
XM_017008126.1 XP_016863615.1 3 - 4 391 127
XM_017008127.1 XP_016863616.1 7 - 8 7691 127
GenBank mRNA AK310262.1 - 5 - 6 647 127

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

transport
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
glutamate receptor signaling pathway
regulation of neuron projection development
cerebellar granule cell differentiation
ion transmembrane transport
cellular protein localization
ionotropic glutamate receptor signaling pathway
synaptic transmission, glutamatergic
regulation of neuron apoptotic process
excitatory postsynaptic potential
prepulse inhibition
ionotropic glutamate receptor activity
extracellular-glutamate-gated ion channel activity
glutamate receptor activity
PDZ domain binding
scaffold protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000502699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000502699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000502699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000502699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000502699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000512631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000512631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000512631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000512631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000512631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000513976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000513976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000513976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000513976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000513976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000515744.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000515744.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000515744.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000515744.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0