This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04974370_m1
See other SGMS2 GE Assays ›
Gene Symbol:
SGMS2
Gene Name:
sphingomyelin synthase 2
Gene Aliases:
SMS2
Chromosome Location:
Chr.4: 107823645 - 107915047 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04974370_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SGMS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136258.1 NP_001129730.1 1 - 2 227 72
XM_011531702.2 XP_011530004.1 1 - 2 1147 72
GenBank mRNA AF452717.1 - 1 - 2 130 72
AK290344.1 - 1 - 2 229 72
DC417367.1 - 1 - 2 234 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF452717.1, AK290344.1, NM_001136258.1, XM_011531702.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

sphingomyelin biosynthetic process
phosphorylation
sphingolipid biosynthetic process
kinase activity
sphingomyelin synthase activity
ceramide cholinephosphotransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000503385.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000503385.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000503385.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000503385.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000503385.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000503862.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000503862.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000503862.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000503862.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000503862.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000506462.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000506462.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000506462.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000506462.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000506462.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000506993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000506993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000506993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000506993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000506993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000515332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000515332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000515332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 ENST00000515332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000359079.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394684.8 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000394686.3 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000503385.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000503385.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000503385.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000503385.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000503385.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000503862.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000503862.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000503862.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000503862.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000503862.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000506462.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000506462.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000506462.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000506462.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000506462.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000506993.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000506993.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000506993.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000506993.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000506993.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000515332.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000515332.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000515332.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 ENST00000515332.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136257.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_001136258.1 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq
Affymetrix Human Clariom D Assay TC0400008345.hg.1 NM_152621.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0