This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04972516_s1
See other CRTAP GE Assays ›
Gene Symbol:
CRTAP
Gene Name:
cartilage associated protein
Gene Aliases:
CASP, LEPREL3, OI7, P3H5
Chromosome Location:
Chr.3: 33113958 - 33147773 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04972516_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRTAP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006371.4 NP_006362.1 7 97
GenBank mRNA AK027235.1 - 1 525 97

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CRTAP

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

spermatogenesis
peptidyl-proline hydroxylation to 3-hydroxy-L-proline
protein stabilization
chaperone-mediated protein folding
negative regulation of post-translational protein modification
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300010667.hg.1 ENST00000607832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010667.hg.1 ENST00000607832.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000485310.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000485310.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000485310.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000485310.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 ENST00000485310.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300010667.hg.1 ENST00000607832.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010667.hg.1 ENST00000607832.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000320954.10 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000449224.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000485310.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000485310.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000485310.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000485310.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 ENST00000485310.1 Ensembl
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq
Affymetrix Human Clariom D Assay TC0300006993.hg.1 NM_006371.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0