This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04971191_m1
See other ALDH1L1 GE Assays ›
Gene Symbol:
ALDH1L1
Gene Name:
aldehyde dehydrogenase 1 family member L1
Gene Aliases:
10-FTHFDH, 10-fTHF, FDH, FTHFD
Chromosome Location:
Chr.3: 126103561 - 126181526 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04971191_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH1L1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA KM111188.1 - 1 - 2 137 96

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the KM111188.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0