This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04970824_m1
See other IGF2BP2-AS1 GE Assays ›
Gene Symbol:
IGF2BP2-AS1
Gene Name:
IGF2BP2 antisense RNA 1
Gene Aliases:
C3orf65
Chromosome Location:
Chr.3: 185713252 - 185729787 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04970824_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGF2BP2-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC039525.1 - 2 - 3 243 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000296270.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000296270.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000296270.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000296270.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000465364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000465364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000470754.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000470754.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000470754.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000496717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000496717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000496717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000496717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000496717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 ENST00000496717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000296270.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000296270.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000296270.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000296270.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000456447.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000465364.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000465364.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000470754.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000470754.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000470754.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000496717.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000496717.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000496717.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000496717.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000496717.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 ENST00000496717.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Clariom D Assay TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Clariom D Assay TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Clariom D Assay TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Clariom D Assay TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Clariom D Assay TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Clariom D Assay TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Clariom D Assay TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Clariom D Assay TC0300009742.hg.1 NR_126326.1 Refseq
Affymetrix Human Clariom D Assay TC0300009742.hg.1 NR_126326.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0