This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04970727_m1
See other HEG1 GE Assays ›
Gene Symbol:
HEG1
Gene Name:
heart development protein with EGF like domains 1
Gene Aliases:
HEG, MST112, MSTP112
Chromosome Location:
Chr.3: 124965710 - 125055958 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04970727_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HEG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005247666.1 XP_005247723.1 5 - 6 1656 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

vasculogenesis
in utero embryonic development
endothelial cell morphogenesis
lymph vessel development
lymph circulation
cardiac atrium morphogenesis
ventricular trabecula myocardium morphogenesis
ventricular septum development
cell-cell junction assembly
post-embryonic development
lung development
multicellular organism growth
venous blood vessel morphogenesis
regulation of body fluid levels
cardiac muscle tissue growth
pericardium development
positive regulation of fibroblast growth factor production
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000477536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000477536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000477536.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000480667.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000480667.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000480667.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000480667.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000482699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000482699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000482699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000482699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000488654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000488654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 ENST00000488654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000311127.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000477536.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000477536.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000477536.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000480667.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000480667.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000480667.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000480667.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000482699.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000482699.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000482699.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000482699.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000487661.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000488654.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000488654.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 ENST00000488654.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq
Affymetrix Human Clariom D Assay TC0300012238.hg.1 NM_020733.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0