This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04967368_s1
See other UCN GE Assays ›
Gene Symbol:
UCN
Gene Name:
urocortin
Gene Aliases:
UI, UROC
Chromosome Location:
Chr.2: 27307400 - 27308445 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04967368_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003353.3 NP_003344.1 2 621 110
GenBank mRNA BC104470.1 - 2 422 40033510 110
BC104471.2 - 2 422 40033515 110
BU619919.1 - 1 110
CR542244.1 - 1 322 110

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UCN

Panther Classification:

Molecular Function -

peptide hormone

Gene Ontology Categories:

Function(s) Process(es)

startle response
inflammatory response
response to oxidative stress
G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
female pregnancy
sensory perception of sound
associative learning
aerobic respiration
negative regulation of gene expression
response to auditory stimulus
pancreatic juice secretion
positive regulation of cell growth
positive regulation of cAMP biosynthetic process
negative regulation of histone deacetylation
neuron projection development
negative regulation of appetite
response to estradiol
positive regulation of interleukin-6 production
positive regulation of collagen biosynthetic process
positive regulation of peptidyl-serine phosphorylation
activation of protein kinase A activity
social behavior
gastric emptying
drinking behavior
negative regulation of apoptotic process
positive regulation of vascular permeability
positive regulation of translation
positive regulation of DNA replication
negative regulation of blood pressure
negative regulation of cell size
positive regulation of vasodilation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of hormone secretion
response to pain
response to glucocorticoid
positive regulation of corticotropin secretion
regulation of synaptic transmission, glutamatergic
positive regulation of cardiac muscle contraction
negative regulation of gastric acid secretion
negative regulation of necrotic cell death
positive regulation of calcium ion import
negative regulation of neuron death
negative regulation of feeding behavior
positive regulation of behavioral fear response
neuropeptide hormone activity
protein binding
corticotropin-releasing hormone activity
histone deacetylase inhibitor activity
corticotropin-releasing hormone receptor 1 binding
corticotropin-releasing hormone receptor 2 binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0