Assay Details
Gene Symbol: SFTPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005264490.4 | XP_005264547.2 | 8 - 9 | 1020 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000473692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000473692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000473692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000473692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000473692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000491167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000491167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000491167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000494165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000494165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000494165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000494165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000393822.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000409383.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000428225.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000473692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000473692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000473692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000473692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000473692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000491167.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000491167.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000491167.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000494165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000494165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000494165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000494165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | ENST00000519937.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_000542.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013286.hg.1 | NM_198843.2 | Refseq |
