This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04965437_m1
See other CNP GE Assays ›
Gene Symbol:
CNP
Gene Name:
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases:
CNP1
Chromosome Location:
Chr.17: 41966741 - 41977736 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04965437_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011524340.2 XP_011522642.1 1 - 2 130 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011524340.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

hydrolase phosphodiesterase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
chemical synaptic transmission
axonogenesis
aging
adult locomotory behavior
cyclic nucleotide catabolic process
response to toxic substance
substantia nigra development
forebrain development
response to lipopolysaccharide
regulation of mitochondrial membrane permeability
oligodendrocyte differentiation
RNA binding
2',3'-cyclic-nucleotide 3'-phosphodiesterase activity
cyclic nucleotide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000441615.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000441615.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000441615.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000441615.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000441615.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000472031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000472031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000472031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000472031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000472031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000472031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000486438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000486438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000486438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000486438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000486438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000486438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000585452.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000585452.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000585452.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000585452.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000585452.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000585452.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000587679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000587679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000587679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000587679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000587679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000589772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000589772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000589772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000589772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000592105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000592105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000592105.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000592446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000592446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000592446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000592446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000592446.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000592861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000592861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000592861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 ENST00000592861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393888.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000393892.7 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000441615.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000441615.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000441615.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000441615.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000441615.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000472031.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000472031.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000472031.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000472031.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000472031.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000472031.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000486438.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000486438.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000486438.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000486438.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000486438.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000486438.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000585452.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000585452.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000585452.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000585452.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000585452.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000585452.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000587679.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000587679.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000587679.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000587679.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000587679.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000589772.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000589772.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000589772.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000589772.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000591945.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000592105.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000592105.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000592105.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000592446.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000592446.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000592446.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000592446.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000592446.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000592861.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000592861.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000592861.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 ENST00000592861.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq
Affymetrix Human Clariom D Assay TC1700007865.hg.1 NM_033133.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0