This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04965399_m1
See other COX10 GE Assays ›
Gene Symbol:
COX10
Gene Name:
COX10, heme A:farnesyltransferase cytochrome c oxidase assembly factor
Gene Aliases:
-
Chromosome Location:
Chr.17: 14069402 - 14208679 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04965399_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COX10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001303.3 NP_001294.2 2 - 3 374 89
GenBank mRNA AK295925.1 - 2 - 3 172 89
AK312718.1 - 2 - 3 269 89
BC000060.2 - 2 - 3 247 3508180 89
BC006394.2 - 2 - 3 247 4136163 89
BT006985.1 - 2 - 3 170 89
U09466.1 - 2 - 3 210 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK295925.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene COX10

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial fission
mitochondrial electron transport, cytochrome c to oxygen
heme biosynthetic process
heme a biosynthetic process
respiratory chain complex IV assembly
aerobic respiration
cellular respiration
heme O biosynthetic process
hydrogen ion transmembrane transport
cytochrome-c oxidase activity
farnesyltranstransferase activity
protoheme IX farnesyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0