This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04962903_m1
See other TTLL8 GE Assays ›
Gene Symbol:
TTLL8
Gene Name:
tubulin tyrosine ligase like 8
Gene Aliases:
-
Chromosome Location:
Chr.22: 50018575 - 50058227 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04962903_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TTLL8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017029176.1 XP_016884665.1 4 - 5 409 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein polyglycylation
ATP binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000477219.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000477219.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009140.hg.1 ENST00000477219.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000266182.10 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000433387.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000477219.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000477219.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009140.hg.1 ENST00000477219.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0