This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942897_s1
See other NKX3-1 GE Assays ›
Gene Symbol:
NKX3-1
Gene Name:
NK3 homeobox 1
Gene Aliases:
BAPX2, NKX3, NKX3.1, NKX3A
Chromosome Location:
Chr.8: 23678693 - 23682937 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942897_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NKX3-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256339.1 NP_001243268.1 3 101
NM_006167.3 NP_006158.2 2 101
NR_046072.1 1 101
GenBank mRNA AF247704.1 - 3 101
BF511129.1 - 1 262 101

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NKX3-1

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

metanephros development
somitogenesis
positive regulation of protein phosphorylation
transcription, DNA-templated
regulation of transcription, DNA-templated
activation of cysteine-type endopeptidase activity involved in apoptotic process
multicellular organism development
salivary gland development
heart development
positive regulation of cell proliferation
negative regulation of cell proliferation
male gonad development
positive regulation of gene expression
negative regulation of gene expression
positive regulation of cell death
positive regulation of phosphatidylinositol 3-kinase signaling
androgen receptor signaling pathway
activation of protein kinase activity
response to testosterone
cellular response to drug
dorsal aorta development
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
protein kinase B signaling
negative regulation of insulin-like growth factor receptor signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of mitotic cell cycle
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
branching morphogenesis of an epithelial tube
negative regulation of epithelial cell proliferation
positive regulation of cell division
pharyngeal system development
branching involved in prostate gland morphogenesis
epithelial cell proliferation involved in salivary gland morphogenesis
negative regulation of epithelial cell proliferation involved in prostate gland development
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to steroid hormone stimulus
cellular response to hypoxia
mitotic cell cycle arrest
negative regulation of estrogen receptor binding
positive regulation of androgen secretion
positive regulation of response to DNA damage stimulus
positive regulation of apoptotic signaling pathway
positive regulation of intrinsic apoptotic signaling pathway
transcription regulatory region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter sequence-specific
core promoter binding
transcription factor activity, sequence-specific DNA binding
androgen receptor activity
protein binding
transcription factor binding
cysteine-type endopeptidase activator activity involved in apoptotic process
estrogen receptor activity
protein kinase activator activity
estrogen receptor binding
histone deacetylase binding
sequence-specific DNA binding
protein self-association
transcription regulatory region DNA binding
MADS box domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0