This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942694_m1
See other LOC100130357 GE Assays ›
Gene Symbol:
LOC100130357
Gene Name:
uncharacterized LOC100130357
Gene Aliases:
-
Chromosome Location:
Chr.6: 13273094 - 13295586 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942694_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100130357
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_134859.1 2 - 3 268 118
GenBank mRNA BX089972.1 - 2 - 3 268 118
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA R99425.1 - 266 Chr. - - [-] on Build 118
GenBankmRNA H67504.1 - 2 - 3 267 Chr. 6 - 13273419 - 13283441 [-] on Build GRCh38 118

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream TBC1D7-LOC100130357 readthrough TBC1D7-LOC100130357 Sense Chr. 6 - 13266542 - 13328583 on NCBI Build GRCh38
Downstream TBC1 domain family member 7 TBC1D7 MGCPH,
PIG51,
TBC7
23.4kb Sense Chr. 6 - 13304951 - 13328583 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000399446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000399446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000399446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000399446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000399446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000399446.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606627.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606627.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000606627.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000612479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000612479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000612479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 ENST00000612479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000399446.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000399446.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000399446.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000399446.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000399446.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000399446.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606627.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606627.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000606627.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000612479.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000612479.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000612479.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 ENST00000612479.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Clariom D Assay TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Clariom D Assay TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Clariom D Assay TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Clariom D Assay TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Clariom D Assay TC0600014240.hg.1 NM_001242698.1 Refseq
Affymetrix Human Clariom D Assay TC0600014240.hg.1 NM_001242698.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0