This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942287_m1
See other FOXP1-AS1 GE Assays ›
Gene Symbol:
FOXP1-AS1
Gene Name:
FOXP1 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 71289758 - 71305854 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942287_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FOXP1-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_126463.1 3 - 4 401 164
GenBank mRNA BC040624.1 - 3 - 4 390 164
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM559480.1 - 401 Chr. - - [-] on Build 164

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream myoD family inhibitor domain-containing protein-like LOC107986096 -983.8kb Anti-sense Chr. 3 - 70194495 - 70312649 on NCBI Build GRCh38
Downstream microRNA 1284 MIR1284 MIRN1284,
hsa-mir-1284,
mir-1284
239.1kb Anti-sense Chr. 3 - 71541970 - 71542089 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300007776.hg.1 ENST00000465742.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007776.hg.1 ENST00000465742.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007776.hg.1 ENST00000465742.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007776.hg.1 ENST00000465742.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007776.hg.1 ENST00000465742.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007776.hg.1 ENST00000465742.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007776.hg.1 NR_126463.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007776.hg.1 NR_126463.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007776.hg.1 NR_126463.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007776.hg.1 NR_126463.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007776.hg.1 NR_126463.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007776.hg.1 NR_126463.1 Refseq
Affymetrix Human Clariom D Assay TC0300007776.hg.1 ENST00000465742.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007776.hg.1 ENST00000465742.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007776.hg.1 ENST00000465742.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007776.hg.1 ENST00000465742.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007776.hg.1 ENST00000465742.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007776.hg.1 ENST00000465742.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007776.hg.1 NR_126463.1 Refseq
Affymetrix Human Clariom D Assay TC0300007776.hg.1 NR_126463.1 Refseq
Affymetrix Human Clariom D Assay TC0300007776.hg.1 NR_126463.1 Refseq
Affymetrix Human Clariom D Assay TC0300007776.hg.1 NR_126463.1 Refseq
Affymetrix Human Clariom D Assay TC0300007776.hg.1 NR_126463.1 Refseq
Affymetrix Human Clariom D Assay TC0300007776.hg.1 NR_126463.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header