This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942268_m1
See other PCAT29 GE Assays ›
Gene Symbol:
PCAT29
Gene Name:
prostate cancer associated transcript 29 (non-protein coding)
Gene Aliases:
-
Chromosome Location:
Chr.15: 69592200 - 69695750 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942268_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCAT29
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_126437.1 1 - 2 117 111
GenBank mRNA AK097902.1 - 1 - 2 190 111
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB106564.1 - 1 - 2 190 Chr. 15 - 69592129 - 69611335 [+] on Build GRCh38 111

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream downregulated RNA in cancer,
inhibitor of cell invasion and migration
DRAIC -20.8kb Sense Chr. 15 - 69561720 - 69571440 on NCBI Build GRCh38
Downstream long intergenic non-protein coding RNA 593 LINC00593 C15orf50 224.2kb Sense Chr. 15 - 69835234 - 69842967 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558411.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000559212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000559212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000559212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000559212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000559212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 NR_126437.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 NR_126437.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 NR_126437.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 NR_126437.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 NR_126437.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 NR_126437.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 NR_126438.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 NR_126438.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 NR_126438.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007702.hg.1 NR_126438.1 Refseq
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558411.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558411.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558411.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558411.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000558941.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000559212.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000559212.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000559212.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000559212.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000559212.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 ENST00000560655.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007702.hg.1 NR_126437.1 Refseq
Affymetrix Human Clariom D Assay TC1500007702.hg.1 NR_126437.1 Refseq
Affymetrix Human Clariom D Assay TC1500007702.hg.1 NR_126437.1 Refseq
Affymetrix Human Clariom D Assay TC1500007702.hg.1 NR_126437.1 Refseq
Affymetrix Human Clariom D Assay TC1500007702.hg.1 NR_126437.1 Refseq
Affymetrix Human Clariom D Assay TC1500007702.hg.1 NR_126437.1 Refseq
Affymetrix Human Clariom D Assay TC1500007702.hg.1 NR_126438.1 Refseq
Affymetrix Human Clariom D Assay TC1500007702.hg.1 NR_126438.1 Refseq
Affymetrix Human Clariom D Assay TC1500007702.hg.1 NR_126438.1 Refseq
Affymetrix Human Clariom D Assay TC1500007702.hg.1 NR_126438.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0