This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04942248_m1
See other ABCA9-AS1 GE Assays ›
Gene Symbol:
ABCA9-AS1
Gene Name:
ABCA9 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 69004714 - 69018323 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04942248_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCA9-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_126414.1 3 - 4 304 144
GenBank mRNA DA128020.1 - 3 - 4 316 144

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ATP binding cassette subfamily A member 8 ABCA8 -56.8kb Anti-sense Chr. 17 - 68867285 - 68955426 on NCBI Build GRCh38
Downstream ATP binding cassette subfamily A member 6 ABCA6 EST155051 43.7kb Anti-sense Chr. 17 - 69062044 - 69142188 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700012300.hg.1 ENST00000458677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012300.hg.1 ENST00000458677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012300.hg.1 ENST00000458677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012300.hg.1 ENST00000458677.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012300.hg.1 NR_126414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012300.hg.1 NR_126414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012300.hg.1 NR_126414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012300.hg.1 NR_126414.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627453.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627453.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627453.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627453.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627453.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000629311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000629311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000629311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000629311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000629311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000630575.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000630575.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000630575.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000630575.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012299.hg.1 ENST00000630575.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627453.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627453.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627453.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627453.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627453.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627596.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627596.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000627957.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000629311.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000629311.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000629311.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000629311.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000629311.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000630575.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000630575.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000630575.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000630575.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012299.hg.1 ENST00000630575.2 Ensembl
Affymetrix Human Clariom D Assay TC1700012300.hg.1 ENST00000458677.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012300.hg.1 ENST00000458677.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012300.hg.1 ENST00000458677.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012300.hg.1 ENST00000458677.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012300.hg.1 NR_126414.1 Refseq
Affymetrix Human Clariom D Assay TC1700012300.hg.1 NR_126414.1 Refseq
Affymetrix Human Clariom D Assay TC1700012300.hg.1 NR_126414.1 Refseq
Affymetrix Human Clariom D Assay TC1700012300.hg.1 NR_126414.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0