Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008839.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163041.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163041.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163041.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163041.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163041.hg.1 |
TC0100008839.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100163037.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163037.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163037.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163037.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163037.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163037.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163037.hg.1 |
TC0100008839.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163048.hg.1 |
TC0100008839.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163045.hg.1 |
TC0100008839.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163035.hg.1 |
TC0100008839.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163044.hg.1 |
TC0100008839.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000418041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443104.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
ENST00000443565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125942.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008839.hg.1 |
NR_125942.1 |
Refseq |