This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04941852_s1
See other MIR378H GE Assays ›
Gene Symbol:
MIR378H
Gene Name:
microRNA 378h
Gene Aliases:
-
Chromosome Location:
Chr.5: 154829458 - 154829540 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04941852_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR378H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_039667.1 1 47 68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA N78271.1 - 1 Chr. 5 - 154829118 - 154829718 [-] on Build GRCh38 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream La ribonucleoprotein domain family member 1 LARP1 LARP -11.9kb Sense Chr. 5 - 154682663 - 154817607 on NCBI Build GRCh38
Downstream CCR4-NOT transcription complex subunit 8 CNOT8 CAF1,
CALIF,
Caf1b,
POP2,
hCAF1
28.4kb Sense Chr. 5 - 154857952 - 154876792 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500009180.hg.1 ENST00000579966.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009180.hg.1 NR_039667.1 Refseq
Affymetrix Human Clariom D Assay TC0500009180.hg.1 ENST00000579966.1 Ensembl
Affymetrix Human Clariom D Assay TC0500009180.hg.1 NR_039667.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0