This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04940023_m1
See other LINC00434 GE Assays ›
Gene Symbol:
LINC00434
Gene Name:
long intergenic non-protein coding RNA 434
Gene Aliases:
-
Chromosome Location:
Chr.13: 60214347 - 60268104 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04940023_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC00434
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_047022.1 1 - 2 151 94
GenBank mRNA BX103562.1 - 1 - 2 151 94

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream diaphanous related formin 3 DIAPH3 AN,
AUNA1,
DIA2,
DRF3,
NSDAN,
diap3,
mDia2
-62.6kb Sense Chr. 13 - 59665583 - 60163985 on NCBI Build GRCh38
Downstream tudor domain containing 3 TDRD3 128.4kb Anti-sense Chr. 13 - 60396457 - 60573879 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6f7678f454-w8g9k:80/100.66.128.142:80.
git-commit: 7b0558793d0e70b0085ba95ba375cdfe261f8cbf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0