This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04939055_g1
See other LOC105371907 GE Assays ›
Gene Symbol:
LOC105371907
Gene Name:
uncharacterized LOC105371907
Gene Aliases:
-
Chromosome Location:
Chr.17: 77273784 - 77281897 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04939055_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC105371907
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_136503.1 2 - 3 441 68
GenBank mRNA CB529354.1 - 1 - 2 68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CD366983.1 - 1 - 2 Chr. 17 - 77273788 - 77281897 [+] on Build GRCh38 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream lung adenocarcinoma-associated transcript 1 LACAT1 Anti-sense Chr. 17 - 77253774 - 77276504 on NCBI Build GRCh38
Downstream septin 9 SEPT9 AF17q25,
MSF,
MSF1,
NAPB,
PNUTL4,
SINT1,
SeptD1
7.1kb Anti-sense Chr. 17 - 77281410 - 77500596 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700011849.hg.1 ENST00000581153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011849.hg.1 ENST00000581153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011849.hg.1 ENST00000581153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011849.hg.1 ENST00000581153.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011849.hg.1 ENST00000581657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011849.hg.1 ENST00000581657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011849.hg.1 ENST00000581657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011849.hg.1 ENST00000581657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011849.hg.1 ENST00000582422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011849.hg.1 ENST00000582422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700011849.hg.1 ENST00000582422.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011849.hg.1 ENST00000581153.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011849.hg.1 ENST00000581153.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011849.hg.1 ENST00000581153.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011849.hg.1 ENST00000581153.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011849.hg.1 ENST00000581657.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011849.hg.1 ENST00000581657.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011849.hg.1 ENST00000581657.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011849.hg.1 ENST00000581657.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011849.hg.1 ENST00000582422.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011849.hg.1 ENST00000582422.1 Ensembl
Affymetrix Human Clariom D Assay TC1700011849.hg.1 ENST00000582422.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0