This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04938837_m1
See other AS-PTPRE GE Assays ›
Gene Symbol:
AS-PTPRE
Gene Name:
lncRNA antisense to PTPRE protein-coding gene
Gene Aliases:
-
Chromosome Location:
Chr.10: 128057732 - 128062425 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04938837_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AS-PTPRE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_136149.1 1 - 2 299 70
NR_136150.1 1 - 2 299 70
GenBank mRNA KR186191.1 - 1 - 2 299 70
KR186192.1 - 1 - 2 299 70
KR186193.1 - 1 - 2 299 70
KR186194.1 - 1 - 2 299 70
KR186195.1 - 1 - 2 207 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream clarin 3 CLRN3 TMEM12,
USH3AL1
-167.7kb Sense Chr. 10 - 127877841 - 127892947 on NCBI Build GRCh38
Downstream marker of proliferation Ki-67 MKI67 KIA,
MIB-,
MIB-1,
PPP1R105
34.2kb Sense Chr. 10 - 128096659 - 128126405 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0