This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04937856_m1
See other LACAT8 GE Assays ›
Gene Symbol:
LACAT8
Gene Name:
lung adenocarcinoma-associated transcript 8
Gene Aliases:
-
Chromosome Location:
Chr.12: 130990626 - 130993976 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04937856_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LACAT8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_131950.1 2 - 3 279 105
GenBank mRNA DB081504.1 - 2 - 3 279 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream adhesion G protein-coupled receptor D1 ADGRD1 GPR133,
PGR25
Anti-sense Chr. 12 - 130953907 - 131141469 on NCBI Build GRCh38
Downstream long intergenic non-protein coding RNA 1257 LINC01257 172.5kb Anti-sense Chr. 12 - 131165011 - 131212931 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 ENST00000536673.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 ENST00000536673.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 ENST00000536673.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 ENST00000542980.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 ENST00000542980.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 ENST00000542980.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 ENST00000542980.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 ENST00000542980.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 NR_131950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 NR_131950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 NR_131950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 NR_131950.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 NR_131951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012460.hg.1 NR_131951.1 Refseq
Affymetrix Human Clariom D Assay TC1200012460.hg.1 ENST00000536673.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012460.hg.1 ENST00000536673.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012460.hg.1 ENST00000536673.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012460.hg.1 ENST00000542980.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012460.hg.1 ENST00000542980.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012460.hg.1 ENST00000542980.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012460.hg.1 ENST00000542980.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012460.hg.1 ENST00000542980.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012460.hg.1 NR_131950.1 Refseq
Affymetrix Human Clariom D Assay TC1200012460.hg.1 NR_131950.1 Refseq
Affymetrix Human Clariom D Assay TC1200012460.hg.1 NR_131950.1 Refseq
Affymetrix Human Clariom D Assay TC1200012460.hg.1 NR_131950.1 Refseq
Affymetrix Human Clariom D Assay TC1200012460.hg.1 NR_131951.1 Refseq
Affymetrix Human Clariom D Assay TC1200012460.hg.1 NR_131951.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0