This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04937817_g1
See other CASP1P2 GE Assays ›
Gene Symbol:
CASP1P2
Gene Name:
caspase 1 pseudogene 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 105062602 - 105071530 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04937817_g1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CASP1P2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_131905.1 3 - 4 580 125

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream caspase recruitment domain family member 16 CARD16 COP,
COP1,
PSEUDO-ICE
-20.1kb Sense Chr. 11 - 105040218 - 105045416 on NCBI Build GRCh38
Downstream caspase recruitment domain family member 17 CARD17 INCA 26.2kb Sense Chr. 11 - 105092469 - 105101487 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100012169.hg.1 ENST00000526345.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012169.hg.1 ENST00000526345.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012169.hg.1 ENST00000526345.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012169.hg.1 ENST00000526345.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header