This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04937816_m1
See other CASP1P2 GE Assays ›
Gene Symbol:
CASP1P2
Gene Name:
caspase 1 pseudogene 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 105062602 - 105071530 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04937816_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CASP1P2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_131905.1 2 - 3 348 105
GenBank mRNA CB985891.1 - 2 - 3 351 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream caspase recruitment domain family member 16 CARD16 COP,
COP1,
PSEUDO-ICE
-20.6kb Sense Chr. 11 - 105040218 - 105045416 on NCBI Build GRCh38
Downstream caspase recruitment domain family member 17 CARD17 INCA 21.6kb Sense Chr. 11 - 105092469 - 105101487 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 ENST00000526830.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq
Affymetrix Human Clariom D Assay TC1100012168.hg.1 NR_131905.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0