This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04937654_g1
See other KCND3-AS1 GE Assays ›
Gene Symbol:
KCND3-AS1
Gene Name:
KCND3 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 111909657 - 111910930 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04937654_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KCND3-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046619.1 3 - 4 304 69
GenBank mRNA AI910896.1 - 1 - 2 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW779128.1 - 1 - 2 Chr. 1 - 111910125 - 111910928 [-] on Build GRCh38 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream KCND3 intronic transcript 1 KCND3-IT1 -53.2kb Anti-sense Chr. 1 - 111853762 - 111856905 on NCBI Build GRCh38
Downstream uncharacterized LOC643355 LOC643355 79.2kb Sense Chr. 1 - 111990100 - 111998842 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000419258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000419258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000419258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000419258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000419258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000419258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000441125.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000441125.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000441125.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000441125.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000441125.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 ENST00000441125.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000419258.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000419258.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000419258.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000419258.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000419258.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000419258.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000427471.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000441125.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000441125.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000441125.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000441125.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000441125.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 ENST00000441125.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Clariom D Assay TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Clariom D Assay TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Clariom D Assay TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Clariom D Assay TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Clariom D Assay TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Clariom D Assay TC0100009431.hg.1 NR_046619.1 Refseq
Affymetrix Human Clariom D Assay TC0100009431.hg.1 NR_046619.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0