This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04935589_m1
See other EGR3 GE Assays ›
Gene Symbol:
EGR3
Gene Name:
early growth response 3
Gene Aliases:
EGR-3, PILOT
Chromosome Location:
Chr.8: 22687661 - 22693302 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04935589_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EGR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199881.1 NP_001186810.1 1 - 2 83 117
GenBank mRNA AK295834.1 - 1 - 2 80 117
DC333411.1 - 1 - 2 83 117

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK295834.1, NM_001199881.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of endothelial cell proliferation
cell migration involved in sprouting angiogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
neuromuscular synaptic transmission
peripheral nervous system development
muscle organ development
circadian rhythm
positive regulation of T cell differentiation in thymus
endothelial cell chemotaxis
cellular response to vascular endothelial growth factor stimulus
negative regulation of apoptotic process
cellular response to fibroblast growth factor stimulus
regulation of gamma-delta T cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000518773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000518773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000524088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000524088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000524088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 ENST00000524088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000317216.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000518773.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000518773.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000519492.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000522910.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000524088.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000524088.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000524088.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 ENST00000524088.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199880.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_001199881.1 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_004430.2 Refseq
Affymetrix Human Clariom D Assay TC0800009856.hg.1 NM_004430.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0